Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0405

Sigma-Aldrich

Lys(Me1)27 Histone H3 Peptide (21-44), biotin

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

assay

≥95% (HPLC)

form

aqueous solution

packaging

pkg of 40 nmol

concentration

40 μM

shipped in

dry ice

storage temp.

−70°C

General description

Histone H3 Peptide, biotin conjugate. The peptide corresponds to Histone H3 residues 21-44 mono-methylated at Lys-27 with an additional C-terminal glycine followed by a biotinylated lysine.

Application

Study enzyme kinetics, and screen small molecular inhibitors of Histone 3 (K27) methyltransferases for drug discovery and HTS applications.

Physical form

Supplied in a TRIS-buffered solution.

Other Notes

1 mL at 40 μM, or 40 nmol Sold as 40 nmol

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service