Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0401

Sigma-Aldrich

Histone H4 peptide (Di-methylated R3), biotin

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
$257.00

$257.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
500 μL
$257.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$257.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥90% (HPLC)

form

aqueous solution

packaging

pkg of 500 μL

concentration

200 μM

shipped in

dry ice

storage temp.

−70°C

General description

Histones have an important role in the organization and modification of chromatin.[1] The nucleosome is the basic unit of the chromatin. It is made up of two molecules each of histones H2A, H2B, H3 and H4 wrapped with approximately 146bp of DNA. Histone H1 binds to the nucleosomes at the DNA entry and exit points and stabilizes them.[2] The gene encoding this protein is localized on human chromosome 6p22.2.[3]

Application

Study enzyme kinetics, and screen small molecular inhibitors of PRMT for drug discovery and HTS applications.

Other Notes

500 μL at 200 μM, or 100 nmol Sold as 500 μL

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Core histones H2B and H4 are mobilized during infection with herpes simplex virus 1.
Conn KL, et al.
Journal of Virology, 85(24), 13234-13252 (2011)
A Comprehensive View of the Epigenetic Landscape Part I: DNA Methylation, Passive and Active DNA Demethylation Pathways and Histone Variants
Anna Sadakierska-Chudy
Neurotoxicity Research, 27, 84-97 (2015)
The histone variant H3.3 marks active chromatin by replication-independent nucleosome assembly.
Ahmad K and Henikoff S
Molecular Cell, 9(6), 1191-1200 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service