Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0389

Sigma-Aldrich

PRMT6 active human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

HRMT1L6, Histone-arginine N-methyltransferase 6, protein arginine N-methyltransferase 6

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$707.00

$707.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
$707.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$707.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

43 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRMT6(55170)

General description

Protein arginine methyltransferase 6 (PRMT6) is a 41.9kDa protein with 375 amino acids. It is encoded by the gene mapped to human chromosome 1. It belongs to type I enzyme, which is a subclass of PRMT family. PRMT6 is localized exclusively in nucleus.[1]
Human PRMT6 (GenBank Accession No. NM_018137), amino-acids 2-375 (end), with N-terminal His-tag, MW=43kDa, expressed in Escherichia coli.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Protein arginine methyltransferase 6 (PRMT6) catalyze the formation of asymmetric ω-NG,NG -dimethylarginine residues. In addition, PRMT6 show automethylation activity, which is unique among the members of PRMT family.[1] PRMT6 mediates cigarette smoke extract (CSE)-triggered apoptosis and inflammation via di-methylation H3 arginine 2 (H3R2me2a) in human umbilical vein endothelial cells (HUVECs).[2] PRMT6 plays a crucial role in various physiological processes such as transcriptional regulation, DNA repair and human immunodeficiency virus pathogenesis.[3] Upregulated expression of the enzyme might be favorable for tumor growth and progression. Overexpression of PRMT6 has been observed in various type of malignancies including breast, cervix, bladder, prostate and lung cancer.[4]

Physical form

Formulated in 200 mM imidazole, 20% glycerol, and 3 mM DTT.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The arginine methyltransferase PRMT6 regulates cell proliferation and senescence through transcriptional repression of tumor suppressor genes.
Stein C
Nucleic Acids Research, 40(19), 9522-9533 (2012)
PRMT6 mediates CSE induced inflammation and apoptosis.
Kang N
International Immunopharmacology, 24(1), 95-101 (2015)
Identification and characterization of new molecular partners for the protein arginine methyltransferase 6 (PRMT6).
Lo Sardo A
PLoS ONE, 8(1), e53750-e53750 (2013)
The novel human protein arginine N-methyltransferase PRMT6 is a nuclear enzyme displaying unique substrate specificity.
Frankel A
The Journal of Biological Chemistry, 277(5), 3537-3543 (2002)
Ernesto Guccione et al.
Nature, 449(7164), 933-937 (2007-09-28)
Eukaryotic genomes are organized into active (euchromatic) and inactive (heterochromatic) chromatin domains. Post-translational modifications of histones (or 'marks') are key in defining these functional states, particularly in promoter regions. Mutual regulatory interactions between these marks--and the enzymes that catalyse them--contribute

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service