Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0350

Sigma-Aldrich

IGF-1R active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)

Synonym(s):

IGF1A, IGFI, insulin-like growth factor 1 (somatomedin C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

72.3 kDa (46 + GST-tag)

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

0.32 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IGF1(3479)

General description

Human IGFR1 (GenBank Accession No. NP_000875) , residues 930-1337 with N-terminal GST-fusion protein containing the 46 kDa cytoplasmic domain of the beta-subunit of the human Insulin-like Growth Factor-1 Receptor Kinase, MW=72.3 kDa, expressed in a Baculovirus expression system.

Application

Enzyme kinetics, autophosphorylation, and phosphorylation of target substrates.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shin-Taek Oh et al.
The American Journal of dermatopathology, 36(10), 807-811 (2014-07-08)
Insulin-like growth factor-1 receptor (IGF-1R) is a key regulator of cell transformation and controls the expression of genes that governs cell cycling and cell survival. The aim of this pilot study was to gain insight into the expression pattern of
N. Tennagels, et al.
Biochemical and Biophysical Research Communications, 260, 7-7 (1999)
K Baer et al.
Biochemistry, 40(47), 14268-14278 (2001-11-21)
To study the role of kinase dimerization in the activation of the insulin receptor (IR) and the insulin-like growth factor receptor-1 (IGF-1R), we have cloned, expressed, and purified monomeric and dimeric forms of the corresponding soluble kinase domains via the
Ying Xia et al.
American journal of translational research, 12(10), 6264-6276 (2020-11-17)
Full-thickness skin injury affects millions of people worldwide each year. Although bone marrow-derived mesenchymal stem cells (BM-MSCs) have been shown to promote cutaneous wound healing, they cannot functionally promote wound healing with the recovery of appendages such as hair follicles.
Michiko Shigyo et al.
Scientific reports, 5, 12055-12055 (2015-07-15)
Vimentin, an intermediate filament protein, is generally recognised as an intracellular protein. Previously, we reported that vimentin was secreted from astrocytes and promoted axonal growth. The effect of extracellular vimentin in neurons was a new finding, but its signalling pathway

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service