Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0338

Sigma-Aldrich

ZAP70 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥50% (SDS-PAGE)

Synonym(s):

SRK, STD, TZK, zeta-chain (TCR) associated protein kinase 70 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥50% (SDS-PAGE)

form

aqueous solution

mol wt

96 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ZAP70(7535)

General description

Recombinant full-length human ZAP70 (GenBank Accession No. NM_001079) full length with N-terminal GST-tag, MW= 96 kDa, was expressed by Baculovirus in Sf9 insect cells.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Fomulated in 50 mM Tris-HCl, pH 7.5,150 mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Pouzolles et al.
The Journal of allergy and clinical immunology, 145(2), 679-697 (2019-09-13)
Patients with T-cell immunodeficiencies are generally treated with allogeneic hematopoietic stem cell transplantation, but alternatives are needed for patients without matched donors. An innovative intrathymic gene therapy approach that directly targets the thymus might improve outcomes. We sought to determine
Mohammad Ishaq et al.
Journal of immunology (Baltimore, Md. : 1950), 179(6), 3851-3858 (2007-09-06)
We have recently shown that Zap70 is important in retinoid receptor-dependent transactivation in T lymphocytes. We report that Zap70 signaling is also essential in dexamethasone-inducible glucocorticoid receptor (GR)-mediated transactivation in T lymphocytes. Zap70-negative Jurkat T cells and cells reconstituted with
Kubra F Naqvi et al.
Frontiers in microbiology, 7, 362-362 (2016-04-06)
The enzymes involved in synthesizing the bacterial cell wall are attractive targets for the design of antibacterial compounds, since this pathway is essential for bacteria and is absent in animals, particularly humans. A survey of the genome of a bacterium
Owen M Siggs et al.
Immunity, 27(6), 912-926 (2007-12-21)
Null mutations that cripple T cell receptor (TCR) signaling explain rare primary immunodeficiencies, but it is not understood why more common polymorphisms that lead to subtle TCR signaling defects are paradoxically associated with autoimmunity. Here we analyzed how a series
Sohye Yoon et al.
PloS one, 10(6), e0126378-e0126378 (2015-06-18)
Adaptive immunity in homeotherms depends greatly on CD4+ Th cells which release cytokines in response to specific antigen stimulation. Whilst bony fish and poikilothermic tetrapods possess cells that express TcR and CD4-related genes (that exist in two forms in teleost

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service