Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0307

Sigma-Aldrich

HDAC Substrate 3, fluorogenic

≥95% (HPLC)

Synonym(s):

HDAC, histone deacetylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

assay

≥95% (HPLC)

form

aqueous solution

packaging

pkg of 250 nmol

shipped in

dry ice

storage temp.

−70°C

General description

A fluorogenic, acetylated peptide substrate for HDAC′s. It is a suitable substrate for many HDAC′s tested so far except class 2a HDAC.

Application

Study enzyme kinetics, and screen small molecule inhibitors for drug discovery and HTS applications.

Physical form

Supplied as a 5 mM solution in DMSO.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service