SRP0301
HDAC6 FLAG tag active human
recombinant, expressed in baculovirus infected Sf9 cells, ≥75% (SDS-PAGE)
Synonym(s):
HD6, JM21, histone deacetylase 6
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
assay
≥75% (SDS-PAGE)
form
aqueous solution
mol wt
132 kDa
packaging
pkg of 5 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Storage temp.
−70°C
Gene Information
human ... HDAC6(10013)
General description
Human HDAC6 (GenBank Accession No. NM_006044), full length with C-terminal Flag tag, MW= 132 kDa, expressed in Baculovirus expression system.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Robert L Strausberg et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(26), 16899-16903 (2002-12-13)
The National Institutes of Health Mammalian Gene Collection (MGC) Program is a multiinstitutional effort to identify and sequence a cDNA clone containing a complete ORF for each human and mouse gene. ESTs were generated from libraries enriched for full-length cDNAs
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service