Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0287

Sigma-Aldrich

Sphingosine kinase 2 (long form) Active human

recombinant, expressed in baculovirus, ≥80% (SDS-PAGE)

Synonym(s):

SK2, SPHK2, SPK 2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$584.00

$584.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$584.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

$584.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus

assay

≥80% (SDS-PAGE)

form

aqueous solution

specific activity

≥20 units/μg

mol wt

74 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SPHK2(56848)

General description

Glucose activates sphingosine kinase 2 in pancreatic β cells leading to a rise in S1P levels, which is important for glucose-stimulated insulin secretion. [1] Human sphingosine kinase 2 (GenBank Accession No.NM_020126), is full length with an N-terminal His tag, has a molecular weight of 74 kDa, and is expressed in a Baculovirus infected Sf9 cell expression system.

Application

Sphingosine kinase 2 is useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling. Sphingosine kinase 2 has been used in a study to assess neuroapoptosis in the developing mouse brain. [2] Sphingosine kinase 2 assays were used to confirm FTY720 bioactivity in a study investigating cranial allograft incorporation. [3]

Unit Definition

One unit of sphingosine kinase activity is defined as the amount of enzyme required to produce 1 pmol of S1P/min

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia Huang et al.
Cell and tissue research, 347(3), 553-566 (2011-08-25)
Endogenous stem cell recruitment to the site of skeletal injury is key to enhanced osseous remodeling and neovascularization. To this end, this study utilized a novel bone allograft coating of poly(lactic-co-glycolic acid) (PLAGA) to sustain the release of FTY720, a
Jamie Cantrell Stanford et al.
The Journal of biological chemistry, 287(16), 13457-13464 (2012-03-06)
Recent studies suggest that sphingolipid metabolism is altered during type 2 diabetes. Increased levels of the sphingolipid ceramide are associated with insulin resistance. However, a role for sphingolipids in pancreatic beta cell function, or insulin production, and release remains to
Goutam Chakraborty et al.
Journal of neurochemistry, 121(5), 806-817 (2012-03-08)
Our previous studies have indicated that de novo ceramide synthesis plays a critical role in ethanol-induced apoptotic neurodegeneration in the 7-day-old mouse brain. In this study, we examined whether the formation of sphingosine 1-phosphate (S1P), a ceramide metabolite, is associated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service