Protein tyrosine phosphatase. Catalytic domain, (amino acids 2091-2490), (Gene bank accession number NM_080685) with N-terminal GST tag, MW = 71.8 kDa, expressed in an E. coli expression system.
Application
Useful for the study of enzyme kinetics, regulation, to dephosphorylate target substrates and for screening inhibitors.
Unit Definition
One unit will hydrolyze 1 nmol p-nitrophenyl phosphate per minute at pH 7.4 and 30°C.
Physical form
Formulation: recombinant proteins were eluted in 25 mM Tris-HCL, 75 mM NaCl, pH 8.0, 0.05% Tween, 10 mM glutathione, 1 mM DTT and 50% glycerol.
Preparation Note
Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.