Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0188

Sigma-Aldrich

Pyruvate kinase M2 Active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

M2-PK tumor, OPA-interacting protein 3 (OIP-3), Pyruvate kinase 3 (PK3), Pyruvate kinase, muscle, Thyroid hormone-binding protein 1 (THBP1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

58.9 kDa

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PKM2(5315)

General description

Human Pyruvate kinase M2 (PKM2) (GenBank Accession No. NM_002654) full length with N-terminal His tag, MW = 58.9 kDa, expressed in an E. coli expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Pyruvate kinase M2 (PKM2) is an isoform of pyruvate kinase that is expressed in certain differentiated cells and in proliferatively active cells. PKM2 is often overexpressed in cancer cells.

Unit Definition

One unit is defined as the amount of enzyme required to transfer a phosphoryl group from phosphoenolpyruvate (PEP) to 1 pmol of ADP/min at 37°C

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 30% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chompoonuch Sawaengsak et al.
International journal of pharmaceutics, 473(1-2), 113-125 (2014-07-08)
An egg-free and broadly effective influenza vaccine that can be produced rapidly, adequately and cost-effectively is needed. In this study, chitosan-associated DNAs prepared at various nitrogen/phosphate charge (N/P) ratios were studied for their physicochemical properties, stability, cytotoxicity, and protein expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service