Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0176

Sigma-Aldrich

SUV39H1 full length Active human

recombinant, expressed in baculovirus infected insect cells

Synonym(s):

Histone-lysine N-methyltransferase, H3 lysine-9 specific 1, KMT1A, Lysine N-methyltransferase 1A, Su(var)3-9 homolog 1, Suppressor of variegation 3-9 homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

form

aqueous solution

mol wt

74 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SUV39H1(6839)

General description

Human SUV39H1 (GenBank Accession No. NM_003173), full length (amino acids 2-end), with N-terminal GST tag, MW = 74 kDa, expressed in Baculovirus infected Sf9 cell expression system.

Unit Definition

One unit is defined as the amount of enzyme required to methylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 130 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service