SRP0167
DNMT3B Active from mouse
recombinant, expressed in baculovirus infected insect cells, ≥30% (SDS-PAGE)
Synonym(s):
DNA (cytosine-5-)-methyltransferase 3b, DNA MTase MmuIIIB
About This Item
biological source
mouse
recombinant
expressed in baculovirus infected insect cells
assay
≥30% (SDS-PAGE)
form
aqueous solution
mol wt
99 kDa
packaging
pkg of 10 μg
concentration
>0.02 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
mouse ... DNMT3B(13436)
General description
Application
Biochem/physiol Actions
Physical form
Preparation Note
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service