Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRE0061

Sigma-Aldrich

Monoclonal Anti-pan-Cytokeratin

clone PCK-26, Reagents designed and manufactured under cGMP controls suitable for use in an IVD application.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PCK-26, monoclonal

contains

15 mM sodium azide

species reactivity

human, chicken, snake, hamster, pig, goat, feline, bovine, carp, rat, rabbit, canine, lizard, sheep, mouse, guinea pig

technique(s)

immunoblotting: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:300
immunohistochemistry (frozen sections): suitable
indirect immunofluorescence: 1:300 using using protease-idgested, formalin-fixed, paraffin-embedded sections of human or animal tissues

isotype

IgG1

shipped in

dry ice

Storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Cytokeratins, a group comprising at least 29 different proteins are characteristic of epithelial and trichocytic cells. Cytokeratin 1, 5, 6, and 8 are members of the type II neutral-to-basic subfamily. Cytokeratin peptide 1 (68 kDa) is expressed together with cytokeratin 10 in the suprabasal cell layers or the differentiation compartment of the epidermis. Its expression increases with epidermal maturation and it is modified post-translationally in the terminally differentiated keratinocytes of the stratum corneum. Cytokeratin peptide 5 (58 kDa) is the primary type II keratin in stratified epithelia while cytokeratin type 8 (52 kDa) is a major type II keratin in simple epithelia. Cytokeratin 6 (56 kDa) is a "hyperproliferation" cytokeratin expressed in tissues with natural or pathological high turnover.

Specificity

The antibody recognizes an epitope located on the Type II cytokeratins 1, 5, 6, and 8. PCK-26 is a broad spectrum antibody which reacts specifically with a variety of normal, reactive, and neoplastic epithelial tissues. The antibody reacts with simple, cornifying, and non-cornifying squamous epithelia and pseudostratified epithelia.

Immunogen

cytokeratin from human epidermis.

Application

Monoclonal antibodies to cytokeratins are specific markers of epithelial cell differentiation and have been widely used as tools in tumor identification and classification. Monoclonal Anti-Cytokeratin, pan is a broadly reactive group-type antibody which recognizes an epitope present in most human epithelial tissues. It facilitates the typing of normal, metaplastic and neoplastic cells and it may aid in the discrimination of carcinomas and non-epithelial tumors such as sarcomas, lymphomas and neural tumors. It is also useful in detecting micro-metastases in lymph nodes and other tissues and for determining the origin of poorly differentiated tumors.
Monoclonal Anti-Cytokeratin, pan may be used for the localization of cytokeratins using various immunochemical assays such as immunoblotting, dot blotting, and immunohistochemstry (immunofluorescence and immunoenzymatic staining).
A minimum antibody titer of 1:300 was determined by indirect immunofluorescent staining of protease digested, formalin-fixed, paraffin-embedded sections of human or animal tissues.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Salomon et al.
Nature communications, 8, 13998-13998 (2017-01-14)
Monolayered epithelia are composed of tight cell assemblies that ensure polarized exchanges. EpCAM, an unconventional epithelial-specific cell adhesion molecule, is assumed to modulate epithelial morphogenesis in animal models, but little is known regarding its cellular functions. Inspired by the characterization
Nayef Abdulrahman Louri et al.
Tissue engineering and regenerative medicine, 19(4), 727-738 (2022-03-25)
In extensive deep dermal burn injuries, split-thickness skin graft (STSG) has been the most preferred treatment option for resurfacing burn wounds. A thick split-thickness skin graft is ideal for preventing graft contracture but is associated with delayed donor healing and
Raffaele Strippoli et al.
Cell death & disease, 11(8), 647-647 (2020-08-20)
Despite their emerging relevance to fully understand disease pathogenesis, we have as yet a poor understanding as to how biomechanical signals are integrated with specific biochemical pathways to determine cell behaviour. Mesothelial-to-mesenchymal transition (MMT) markers colocalized with TGF-β1-dependent signaling and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service