Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2798

Sigma-Aldrich

CH625

≥98% (HPLC)

Synonym(s):

(2E)-1-(4-Hydroxy-2,6-dimethoxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one, 4,4′-Dihydroxy-2′,6′-dimethoxychalcone, ZINC95910725

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$62.60
25 MG
$268.00

$62.60

List Price$65.90Save 5%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$62.60
25 MG
$268.00

About This Item

Empirical Formula (Hill Notation):
C17H16O5
CAS Number:
Molecular Weight:
300.31
UNSPSC Code:
51111800
NACRES:
NA.77

$62.60

List Price$65.90Save 5%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

Storage temp.

2-8°C

Biochem/physiol Actions

CH625 is a potent and selective competitive inhibitor of CYP4X1 that decreases tumor burden, increases survival in glioma intracranial and subcutaneous model in rats. CH625 normalizes tumor vasculature in the C6 and GL261 gliomas through tumor-associated macrophages (TAMs) reprograming and inhibition of VEGF and TGF-β production.
potent and selective competitive inhibitor of CYP4X1 that decreases tumor burden, increases survival in gliomas

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenlong Wang et al.
The Journal of pharmacology and experimental therapeutics, 365(1), 72-83 (2018-02-14)
Tumor-associated macrophages (TAMs) are pivotal effector cells in angiogenesis. Here, we tested whether CYP4X1 inhibition in TAMs by flavonoid CH625 prolongs survival and normalizes glioma vasculature. CH625 was selected against the CYP4X1 3D model by virtual screening and showed inhibitory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service