Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2680

Sigma-Aldrich

ES24

≥98% (HPLC)

Synonym(s):

2-{2-[3-(5-nitro-2-furyl)-2-propen-1-ylidene]hydrazino}-2-oxoethylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$80.75
25 MG
$276.00

$80.75

List Price$85.00Save 5%
Web-Only Promotion

Available to ship onMay 02, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$80.75
25 MG
$276.00

About This Item

Empirical Formula (Hill Notation):
C15H13N3O4
Molecular Weight:
299.28
UNSPSC Code:
12352200
NACRES:
NA.77

$80.75

List Price$85.00Save 5%
Web-Only Promotion

Available to ship onMay 02, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

Quality Level

assay

≥98% (HPLC)

form

powder

color

faint yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

ES24 is an Eeyarestatin 1 analog that blocks protein translocation in vitro and more effectively enhances Sec61-mediated Ca2+ leakage from Endoplasmic Reticulum (ER). Apparently ES24 keep Sec61 in open position allowing Ca2+ ions to move freely through the aqueous channel present. It causes cell-type-specific Ca2+-dependent cell death.
blocks protein translocation in vitro and effectively enhances Sec61-mediated Ca2+ leakage from Endoplasmic Reticulum (ER).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor Gamayun et al.
Cell chemical biology, 26(4), 571-583 (2019-02-26)
Eeyarestatin 1 (ES1) inhibits p97-dependent protein degradation, Sec61-dependent protein translocation into the endoplasmic reticulum (ER), and vesicular transport within the endomembrane system. Here, we show that ES1 impairs Ca2+ homeostasis by enhancing the Ca2+ leakage from mammalian ER. A comparison

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service