Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2582

Sigma-Aldrich

LDK1305

≥98% (HPLC)

Synonym(s):

1-(4-Cyanophenyl)-3-(3-(6-(pyrrolidin-1-yl)pyridin-2-yl)phenyl)urea, N-(4-Cyanophenyl)-N′-[3-[6-(1-pyrrolidinyl)-2-pyridinyl]phenyl]-urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$81.60
25 MG
$242.20

$81.60

List Price$85.90Save 5%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$81.60
25 MG
$242.20

About This Item

Empirical Formula (Hill Notation):
C23H21N5O
CAS Number:
Molecular Weight:
383.45
UNSPSC Code:
12352200
NACRES:
NA.77

$81.60

List Price$85.90Save 5%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(C=C1)C#N)NC2=CC(C3=CC=CC(N4CCCC4)=N3)=CC=C2

InChI

1S/C23H21N5O/c24-16-17-9-11-19(12-10-17)25-23(29)26-20-6-3-5-18(15-20)21-7-4-8-22(27-21)28-13-1-2-14-28/h3-12,15H,1-2,13-14H2,(H2,25,26,29)

InChI key

HWIMBWAXFAEZQR-UHFFFAOYSA-N

Biochem/physiol Actions

LDK1305 is a selective positive allosteric modulator of CB1 that promote the binding of the agonist CP55940 and inhibits binding of orthosteric inverse agonist SR141716A. LDK1305 is a beta-arrestin-biased ligand that induces cellular internalization of the receptor and recruitment of β-arrestin 2 in HEK293 cells. It stimulates also β-arrestin 1-dependent phosphorylation of the downstream kinases, ERK1/2, MEK, and Src.
selective positive allosteric modulator of CB1; a beta-arrestin-biased ligand

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service