Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2562

Sigma-Aldrich

Raphin1

≥98% (HPLC)

Synonym(s):

2-[(2,3-Dichlorophenyl)methylene]hydrazinecarboximidamide, N-(2, 3-Dichlorobenzylideneamino) guanidine, Rational inhibitor of a holophosphatase1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$85.00
25 MG
$345.00

$85.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$85.00
25 MG
$345.00

About This Item

Empirical Formula (Hill Notation):
C8H8Cl2N4
CAS Number:
Molecular Weight:
231.08
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$85.00


Check Cart for Availability

Request a Bulk Order

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

Clc1c(cccc1\C=N\N=C(N)N)Cl

InChI

1S/C8H8Cl2N4/c9-6-3-1-2-5(7(6)10)4-13-14-8(11)12/h1-4H,(H4,11,12,14)/b13-4+

InChI key

WLTSTDGGFCQWTK-YIXHJXPBSA-N

Biochem/physiol Actions

Raphin1 is an orally available, brain penetrant, potent and selective inhibitor of protein phosphatase 1 regulatory subunit 15B (PPP1R15B; R15B). It does not inhibit PP1c. Raphin1 transiently decreases protein synthesis in cells. It is efficacious in mouse model of Huntington′s disease.
orally available, brain penetrant, potent and selective inhibitor of protein phosphatase 1 regulatory subunit 15B (PPP1R15B; R15B).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service