Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2552

Sigma-Aldrich

AP-C5

≥98% (HPLC)

Synonym(s):

4-[4-(1H-Imidazol-1-yl)phenyl]-N-2-propyn-1-yl-2-pyrimidinamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$60.58
25 MG
$207.00

$60.58

List Price$63.90Save 5%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$60.58
25 MG
$207.00

About This Item

Empirical Formula (Hill Notation):
C16H13N5
CAS Number:
Molecular Weight:
275.31
UNSPSC Code:
12352200
NACRES:
NA.77

$60.58

List Price$63.90Save 5%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C#CCNC1=NC=CC(C2=CC=C(N3C=NC=C3)C=C2)=N1

InChI

1S/C16H13N5/c1-2-8-18-16-19-9-7-15(20-16)13-3-5-14(6-4-13)21-11-10-17-12-21/h1,3-7,9-12H,8H2,(H,18,19,20)

InChI key

QKHQFSQJQKGFAA-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

AP-C5 is a potent and selective inhibitor of the guanosine 3′,5′-cyclicmonophosphate (cGMP)-dependent protein kinase II (cGKII)that selectively blocks cGKII dependent phosphorylation of the vasodilator-stimulated phosphoprotein (VASP) in intestinal organoids. AP-C5 mitigates secretory response provoked by the GCC-activating Escherichia coli heat-stable toxin (STa) in mouse small intestinal tissue.
potent and selective inhibitor of the guanosine 3′,5′-cyclicmonophosphate (cGMP)-dependent protein kinase II (cGKII)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcel J C Bijvelds et al.
The Journal of biological chemistry, 293(21), 8173-8181 (2018-04-15)
The guanosine 3',5'-cyclic monophosphate (cGMP)-dependent protein kinase II (cGKII) serine/threonine kinase relays signaling through guanylyl cyclase C (GCC) to control intestinal fluid homeostasis. Here, we report the discovery of small-molecule inhibitors of cGKII. These inhibitors were imidazole-aminopyrimidines, which blocked recombinant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service