Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2507

Sigma-Aldrich

Cefotetan acid

Synonym(s):

(6R,7S)-7-[[[4-(2-Amino-1-carboxy-2-oxoethylidene)-1,3-dithietan-2-yl]carbonyl]amino]-7-methoxy-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N7O8S4
CAS Number:
Molecular Weight:
575.62
UNSPSC Code:
12352200

form

powder

storage temp.

−20°C

InChI

1S/C17H17N7O8S4/c1-23-16(20-21-22-23)34-4-5-3-33-15-17(32-2,14(31)24(15)7(5)11(29)30)19-9(26)13-35-12(36-13)6(8(18)25)10(27)28/h13,15H,3-4H2,1-2H3,(H2,18,25)(H,19,26)(H,27,28)(H,29,30)/b12-6-/t13?,15-,17+/m1/s1

InChI key

SRZNHPXWXCNNDU-IXOPCIAXSA-N

Biochem/physiol Actions

Cefotetan is a semi-synthetic, broad-spectrum, beta-lactamase-resistant, second-generation cephalosporin antibiotic derived from cephalosporium. The bactericidal activity of cefotetan disodium is caused by an inhibition of bacterial cell wall synthesis via inhibition of cross-linking of peptidoglycans. This results in a reduction of cell wall stability and causes cell lysis. Cefotetan disodium is more active against gram-negative organisms and less active against gram-positive organisms compared to first-generation cephalosporins.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1B

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service