Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2505

Sigma-Aldrich

PRT-060318

≥95% (HPLC)

Synonym(s):

2-((1R,2S)-2-Aminocyclohexylamino)-4-(m-tolylamino)pyrimidine-5-carboxamide, 2-[[(1R,2S)-2-Aminocyclohexyl]amino]-4-[(3-methylphenyl)amino]-5-pyrimidinecarboxamide, P142-76, PRT060318, PRT318

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$96.30
25 MG
$303.00

$96.30

List Price$107.00Save 10%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$96.30
25 MG
$303.00

About This Item

Empirical Formula (Hill Notation):
C18H24N6O
CAS Number:
Molecular Weight:
340.42
MDL number:
UNSPSC Code:
12352200

$96.30

List Price$107.00Save 10%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N[C@H]1CCCC[C@H]1NC2=NC(NC3=CC=CC(C)=C3)=C(C=N2)C(N)=O

InChI key

NZNTWOVDIXCHHS-LSDHHAIUSA-N

Biochem/physiol Actions

Orally active, highly potent and selective Syk inhibitor with in vivo efficacy in animal models of thrombosis and B-cell acute lymphoblastic leukemia (B-ALL).
PRT-060318 (PRT318; P142-76) is a highly potent and selective Syk inhibitor (IC50 = 4 nM; much reduced potency against 138 other kinases) that blocks BCR-dependent signaling in chronic lymphocytic leukemia (CCL) cultures (2-3 μM) and specifically inhibits platelets activation via ITAM receptor complex GPVI/FcRγ, but not P2Y1/12 or PAR1, stimulation (IC50 = 2.5 μM; aggregation by 8 ng/mL convulxin). PRT318 demonstrates in vivo efficacy in animal models of thrombosis (30 mg/kg mouse bis p.o.; 5.1675 mg/3.1 mL/kg/rabbit/15 min then 7.33 mg/4.4 mL/kg/h i.v.; 8.90 mg/mL/kg pig/h i.v.) and B-cell acute lymphoblastic leukemia (30 mg /kg bis p.o.; human B-ALL xenograft mice).

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Self-react. C

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachit Badolia et al.
International journal of molecular sciences, 18(6) (2017-06-10)
The binding of von Willebrand factor (VWF) to the platelet membrane glycoprotein 1b-IX (GP1b-IX) leads to activation of platelets. GP1b was shown to signal via the FcRγ-ITAM (Fc Receptor γ-Immunoreceptor tyrosine-based activation motif) pathway, activating spleen tyrosine kinase (Syk) and
Pierre H Mangin et al.
Haematologica, 103(5), 898-907 (2018-02-24)
Glycoprotein VI, a major platelet activation receptor for collagen and fibrin, is considered a particularly promising, safe antithrombotic target. In this study, we show that human glycoprotein VI signals upon platelet adhesion to fibrinogen. Full spreading of human platelets on
Callum N Watson et al.
Platelets, 27(6), 535-540 (2016-03-31)
Gram-negative Escherichia coli cause diseases such as sepsis and hemolytic uremic syndrome in which thrombotic disorders can be found. Direct platelet-bacterium interactions might contribute to some of these conditions; however, mechanisms of human platelet activation by E. coli leading to
Michael P Reilly et al.
Blood, 117(7), 2241-2246 (2010-11-23)
Heparin-induced thrombocytopenia (HIT) is a major cause of morbidity and mortality resulting from the associated thrombosis. Extensive studies using our transgenic mouse model of HIT have shown that antibodies reactive with heparin-platelet factor 4 complexes lead to FcγRIIA-mediated platelet activation
J Hoellenriegel et al.
Leukemia, 26(7), 1576-1583 (2012-03-01)
Syk is a protein tyrosine kinase that couples B-cell receptor (BCR) activation with downstream signaling pathways, affecting cell survival and proliferation. Moreover, Syk is involved in BCR-independent functions, such as B-cell migration and adhesion. In chronic lymphocytic leukemia (CLL), Syk

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service