Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2477

Sigma-Aldrich

Squarunkin A hydrochloride

≥98% (HPLC)

Synonym(s):

4-[[3,4-Dioxo-2-[[2-[4-[3-(trifluoromethyl)phenyl]-1-piperazinyl]ethyl]amino]-1-cyclobuten-1-yl]amino]-1-piperidinecarboxylic acid ethyl ester, hydrochloride, Ethyl 4-(3,4-dioxo-2-(2-(4-(3-(trifluoromethyl)phenyl)piperazin-1-yl)ethylamino)cyclobut-1-enylamino)piperidine-1-carboxylate, hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$80.75
25 MG
$276.00

$80.75

List Price$85.00Save 5%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$80.75
25 MG
$276.00

About This Item

Empirical Formula (Hill Notation):
C25H32F3N5O4 · xHCl
CAS Number:
Molecular Weight:
523.55 (free base basis)
UNSPSC Code:
12352200

$80.75

List Price$85.00Save 5%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

Storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)N2CCN(CC2)CCNC3=C(C(=O)C3=O)NC4CCN(CC4)C(=O)OCC

InChI

1S/C25H32F3N5O4/c1-2-37-24(36)33-9-6-18(7-10-33)30-21-20(22(34)23(21)35)29-8-11-31-12-14-32(15-13-31)19-5-3-4-17(16-19)25(26,27)28/h3-5,16,18,29-30H,2,6-15H2,1H3

Inchi Key

ZKITWOVRRSBKFG-UHFFFAOYSA-N

Biochem/physiol Actions

Non-cytotoxic, potent and specific UNC119-cargo interation blocker that inhibits Lck localization at T-cell immune synpase and Src activation.
Squarunkin A is a non-cytotoxic, potent and specific UNC119-cargo interation blocker (IC50 = 10 nM against 800 nM myristoylated Src N-terminal peptide for GST-UNC119A) that targets UNC119A/B myristoyl-binding pocket without affecting the interaction of geranylgeranylated Rab1 to RhoGDI22, or farnesylated Rheb to PDE6d, AIPL1, and calmodulin. Squarunkin A blocks Lck localization at staphylococcal E enterotoxin-induced Jurkat T-cell synpase (2 μM) and prevents MDA-MB-231 cellular Src activation (37/62% reduced Src pY416 post 24 hr 78/625 nM Squarunkin A incubation) without growth inhibition or apoptosis induction (10 μM for >35 hrs).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tom Mejuch et al.
Angewandte Chemie (International ed. in English), 56(22), 6181-6186 (2017-05-05)
N-Terminal myristoylation facilitates membrane binding and activity of proteins, in particular of Src family kinases, but the underlying mechanisms are only beginning to be understood. The chaperones UNC119A/B regulate the cellular distribution and signaling of N-myristoylated proteins. Selective small-molecule modulators
Nelli Erwin et al.
Chembiochem : a European journal of chemical biology, 19(14), 1482-1487 (2018-04-28)
Plasma membrane localization of myristoylated c-Src, a proto-oncogene protein-tyrosine kinase, is required for its signaling activity. Recent studies proposed that UNC119 protein functions as a solubilizing factor for myristoylated proteins, thereby regulating their subcellular distribution and signaling. The underlying molecular
Louise A Stephen et al.
Developmental cell, 47(1), 122-132 (2018-09-18)
Upon engagement of the T cell receptor with an antigen-presenting cell, LCK initiates TCR signaling by phosphorylating its activation motifs. However, the mechanism of LCK activation specifically at the immune synapse is a major question. We show that phosphorylation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service