Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2447

Sigma-Aldrich

TPA023

≥98% (HPLC)

Synonym(s):

7-(1,1-Dimethylethyl)-6-[(1-ethyl-1H-1,2,4-triazol-5-yl)methoxy]-3-(2-fluorophenyl)-1,2,4-triazolo[4,3-b]pyridazine, TPA 023

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$86.00
25 MG
$325.00

$86.00

List Price$100.00Save 14%
Web-Only Promotion

Check Cart for Availability


Select a Size

Change View
5 MG
$86.00
25 MG
$325.00

About This Item

Empirical Formula (Hill Notation):
C20H22FN7O
CAS Number:
Molecular Weight:
395.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$86.00

List Price$100.00Save 14%
Web-Only Promotion

Check Cart for Availability

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(C)(C)C1=CC2=NN=C(C3=CC=CC=C3F)N2N=C1OCC4=NC=NN4CC

InChI

1S/C20H22FN7O/c1-5-27-17(22-12-23-27)11-29-19-14(20(2,3)4)10-16-24-25-18(28(16)26-19)13-8-6-7-9-15(13)21/h6-10,12H,5,11H2,1-4H3

InChI key

QKIWQBLNTSQOLY-UHFFFAOYSA-N

Biochem/physiol Actions

Highly potent alpha2/alph3 GABA-A receptor partial agonist for benzodiazepine binding site with anxiolytic activity but lacking sedative activity
TPA023 is a highly potent, functionally selective agonist for α2-, α3-, and α5GABAA receptors that shows zero efficacy at a1GABAA subtypes. TPA023 exhibits anxiolytic and anticonvulsant effects in both animals and humans but lacks sedative effects and abuse potential.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina M Shinday et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 38(6), 1006-1014 (2013-01-11)
Benzodiazepines are prescribed widely but their utility is limited by unwanted side effects, including abuse potential. The mechanisms underlying the abuse-related effects of benzodiazepines are not well understood, although α1 subunit-containing GABAA receptors have been proposed to have a critical
John R Atack et al.
The Journal of pharmacology and experimental therapeutics, 332(1), 17-25 (2009-09-26)
The GABA(A) receptor alpha2/alpha3 subtype-selective compound 7-(1,1-dimethylethyl)-6-(2-ethyl-2H-1,2,4-triazol-3-ylmethoxy)-3-(2-fluorophenyl)-1,2,4-triazolo[4,3-b]pyridazine (TPA023; also known as MK-0777) is a triazolopyridazine that has similar, subnanomolar affinity for the benzodiazepine binding site of alpha1-, alpha2-, alpha3-, and alpha5-containing GABA(A) receptors and has partial agonist efficacy at the
Angela N Duke et al.
The Journal of pharmacology and experimental therapeutics, 366(1), 145-157 (2018-05-04)
In nonhuman primates we tested a new set of behavioral categories for observable sedative effects using pediatric anesthesiology classifications as a basis. Using quantitative behavioral observation techniques in rhesus monkeys, we examined the effects of alprazolam and diazepam (nonselective benzodiazepines)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service