Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2374

Sigma-Aldrich

HTS01037

≥98% (HPLC)

Synonym(s):

(E)-4-(5-(Methoxycarbonyl)-2,2′-bithiophen-4-ylamino)-4-oxobut-2-enoic acid, 4-[(3-Carboxy-1-oxo-2-propenyl)amino]-[2,2′-bithiophene]-5-carboxylic acid 5-methyl ester, 4-{[2-(Methoxycarbonyl)-5-(2-thienyl)-3-thienyl]amino}-4-oxo-2-butenoic acid, HTS 01037, HTS-01037

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$63.84
25 MG
$271.00

$63.84

List Price$67.20
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$63.84
25 MG
$271.00

About This Item

Empirical Formula (Hill Notation):
C14H11NO5S2
CAS Number:
Molecular Weight:
337.37
MDL number:
UNSPSC Code:
12352200

$63.84

List Price$67.20
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1c(cc(c1C(=O)OC)NC(=O)C=CC(=O)O)c2[s]ccc2

InChI

1S/C14H11NO5S2/c1-20-14(19)13-8(15-11(16)4-5-12(17)18)7-10(22-13)9-3-2-6-21-9/h2-7H,1H3,(H,15,16)(H,17,18)

InChI key

GJODSFZNKNHKML-UHFFFAOYSA-N

Biochem/physiol Actions

HTS01037 is a selective, high-affinity fatty acid-binding protein FABP4 (AFABP; aP2) antagonist (aP2 Ki = 0.67 μM; EFABP/HF/IFABP/LFABP Ki = 3.40/9.07/6.57/8.17 μM) that targets aP2 long-chain fatty acid-binding site. HTS01037 is shown to downregulate basal and fatty acid-stimulated LTC4 levels in primary murine peritoneal macrophages (0.2-20 μM), upregulate murine macrophage RAW 264.7 intracellular free fatty acids and uncoupling protein 2 (UCP2) mRNA levels (30 μM), as well as suppress PPARγ target genes expression in IL-4 polarized human primary macrophages (10-25 μM).
Selective, high-affinity fatty acid-binding protein FABP4 (AFABP; aP2) antagonist that blocks aP2-mediated responses in human and murine macrophages.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaylee A Steen et al.
Molecular and cellular biology, 37(2) (2016-11-01)
Obesity-linked metabolic disease is mechanistically associated with the accumulation of proinflammatory macrophages in adipose tissue, leading to increased reactive oxygen species (ROS) production and chronic low-grade inflammation. Previous work has demonstrated that deletion of the adipocyte fatty acid-binding protein (FABP4/aP2)
Eric K Long et al.
Biochimica et biophysica acta, 1831(7), 1199-1207 (2013-04-16)
Obesity results in increased macrophage recruitment to adipose tissue that promotes a chronic low-grade inflammatory state linked to increased fatty acid efflux from adipocytes. Activated macrophages produce a variety of pro-inflammatory lipids such as leukotriene C4 (LTC4) and 5-, 12-
Yuta Okamura et al.
Pflugers Archiv : European journal of physiology, 469(9), 1177-1188 (2017-04-14)
Fatty acid-binding protein (FABP) 4 is an adipocytokine mainly expressed in adipocyte and macrophage. Blood FABP4 is related not only to metabolic disorders including insulin resistance and atherosclerosis but also increased blood pressure. We tested the hypothesis that FABP4 plays
Kuok Teong Ong et al.
Journal of lipid research, 55(5), 808-815 (2014-03-13)
Adipose TG lipase (ATGL) catalyzes the rate-limiting step in TG hydrolysis in most tissues. We have shown that hepatic ATGL preferentially channels hydrolyzed FAs to β-oxidation and induces PPAR-α signaling. Previous studies have suggested that liver FA binding protein (L-FABP)
Hongliang Xu et al.
Molecular and cellular biology, 35(6), 1055-1065 (2015-01-15)
Chronic inflammation in obese adipose tissue is linked to endoplasmic reticulum (ER) stress and systemic insulin resistance. Targeted deletion of the murine fatty acid binding protein (FABP4/aP2) uncouples obesity from inflammation although the mechanism underlying this finding has remained enigmatic.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service