Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2329

Sigma-Aldrich

RP22

≥98% (HPLC)

Synonym(s):

AA147 Inactive Control, AA147 Negative Control, N-(2-Hydroxy-5-(trifluoromethyl)phenyl)-3-phenylpropanamide, N-[2-hydroxy-5-(trifluoromethyl)phenyl]-benzenepropanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3NO2
CAS Number:
Molecular Weight:
309.28
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(C(F)(F)F)=CC=C1O)CCC2=CC=CC=C2

Biochem/physiol Actions

Inactive analog of the ATF6α-selective activator AA147.
RP22 is a AA147 analog with no ATF6α-activating potency. RP22 is the recommended inactive control for AA147 in probing ATF6α signaling-mediated unfolded protein response (UPR).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heike Kroeger et al.
Science signaling, 11(517) (2018-02-15)
ATF6 encodes a transcription factor that is anchored in the endoplasmic reticulum (ER) and activated during the unfolded protein response (UPR) to protect cells from ER stress. Deletion of the isoform activating transcription factor 6α (ATF6α) and its paralog ATF6β

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service