Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2303

Sigma-Aldrich

Tosedostat

≥98% (HPLC)

Synonym(s):

2S-[2R-(S-Hydroxy-hydroxycarbamoyl-methyl)-4-methylpentanoyl-amino]-2-phenylethanoic acid cyclopentyl ester, CHR-2797

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30N2O6
CAS Number:
Molecular Weight:
406.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(C)C[C@H]([C@H](O)C(NO)=O)C(N[C@@H](C1=CC=CC=C1)C(OC2CCCC2)=O)=O

InChI

1S/C21H30N2O6/c1-13(2)12-16(18(24)20(26)23-28)19(25)22-17(14-8-4-3-5-9-14)21(27)29-15-10-6-7-11-15/h3-5,8-9,13,15-18,24,28H,6-7,10-12H2,1-2H3,(H,22,25)(H,23,26)/t16-,17+,18+/m1/s1

Inchi Key

FWFGIHPGRQZWIW-SQNIBIBYSA-N

Biochem/physiol Actions

Potent cell permeable, antiproliferative aminopeptidase inhibitor
Tosedostat (CHR-2797) is a potent cell permeable, antiproliferative aminopeptidase inhibitor. It has been in clinical trials for AML, pancreatic cancer and more.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caner Saygin et al.
Journal of hematology & oncology, 10(1), 93-93 (2017-04-20)
Acute myeloid leukemia (AML) is characterized by clinical and biological heterogeneity. Despite the advances in our understanding of its pathobiology, the chemotherapy-directed management has remained largely unchanged in the past 40 years. However, various novel agents have demonstrated clinical activity, either
Danielle Shafer et al.
Blood reviews, 30(4), 275-283 (2016-03-15)
Acute myeloid leukemia (AML) remains a challenge to both patients and clinicians. Despite improvements in our understanding of the disease, treatment has changed minimally and outcomes remain poor for the majority of patients. Within the last decade, there have been
Raya Mawad et al.
British journal of haematology, 172(2), 238-245 (2015-11-17)
Tosedostat, an oral aminopeptidase inhibitor, has synergy with cytarabine and hypomethylating agents. We performed a Phase II trial to determine rates of complete remission (CR) and survival using tosedostat with cytarabine or decitabine in older patients with untreated acute myeloid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service