Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2222

Sigma-Aldrich

Carisbamate

≥98% (HPLC)

Synonym(s):

(S)-2-O-Carbamoyl-1-o-chlorophenyl-ethanol, JNJ-10234094, RWJ-333369, YKP 509, [(2S)-2-(2-chlorophenyl)-2-hydroxyethyl] carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$106.40

$106.40

List Price$112.00
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
$106.40

About This Item

Empirical Formula (Hill Notation):
C9H10ClNO3
CAS Number:
Molecular Weight:
215.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$106.40

List Price$112.00
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +77 to +87°, c = 0.2 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=C([C@H](O)COC(N)=O)C=CC=C1

InChI

1S/C9H10ClNO3/c10-7-4-2-1-3-6(7)8(12)5-14-9(11)13/h1-4,8,12H,5H2,(H2,11,13)/t8-/m1/s1

InChI key

OLBWFRRUHYQABZ-MRVPVSSYSA-N

Biochem/physiol Actions

Antiepileptic; broad acute anticonvulsant, neuroprotective.
Carisbamate is an antiepileptic agent with broad anticonvulsant and neuroprotective activity. It has also been investigated as a possible treatment for alcohol dependence.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Eduardo Marques-Carneiro et al.
Pharmaceuticals (Basel, Switzerland), 10(4) (2017-11-07)
The administration of lithium-pilocarpine (LiPilo) in adult rats is a validated model reproducing the main clinical and neuropathological features of temporal lobe epilepsy (TLE). Previous studies have shown that carisbamate (CRS) has the property of modifying epileptogenesis in this model.
Do Young Kim et al.
Epilepsia, 58(4), 617-626 (2017-02-24)
Carisbamate (CRS) is a novel monocarbamate compound that possesses antiseizure and neuroprotective properties. However, the mechanisms underlying these actions remain unclear. Here, we tested both direct and indirect effects of CRS on several cellular systems that regulate intracellular calcium concentration
Erwin Z Mangubat et al.
Journal of neurosurgery, 122(6), 1283-1292 (2015-02-28)
The authors evaluated the preclinical feasibility of acutely stabilizing an active bihemispheric limbic epileptic circuit using closed-loop direct neurostimulation therapy in tandem with "on-demand'" convection-enhanced intracerebral delivery of the antiepileptic drug (AED) carisbamate. A rat model of electrically induced self-sustained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service