Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2186

Sigma-Aldrich

Poloppin

≥98% (HPLC)

Synonym(s):

5-(4-Bromophenyl)-1-[2-(trifluoromethyl)phenyl]-1H-pyrrole-2-propanoic acid, Polo protein-protein interaction inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$67.19
25 MG
$360.00

$67.19

List Price$71.10Save 5%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$67.19
25 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C20H15BrF3NO2
CAS Number:
Molecular Weight:
438.24
UNSPSC Code:
12352200
NACRES:
NA.77

$67.19

List Price$71.10Save 5%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

Storage temp.

2-8°C

SMILES string

OC(CCC1=CC=C(C2=CC=C(Br)C=C2)N1C3=CC=CC=C3C(F)(F)F)=O

InChI

1S/C20H15BrF3NO2/c21-14-7-5-13(6-8-14)17-11-9-15(10-12-19(26)27)25(17)18-4-2-1-3-16(18)20(22,23)24/h1-9,11H,10,12H2,(H,26,27)

Inchi Key

OCUKOJUZSNFVFQ-UHFFFAOYSA-N

Biochem/physiol Actions

Poloppin is a cell penetrant and potent inhibitor of mitotic Polo-like kinases (PLKs) that binds to the Polo-box domain (PBD) and prevents PLKs interaction with pSer/pThr phosphopeptide substrates. Poloppin induces mitotic arrest and apoptosis in cancer cells expressing mutant KRAS in 2D and organoid cultures. It also sensitizes mutant KRAS-expressing cancer cells to Crizotinib.
cell penetrant and potent inhibitor of PLKs that binds to the Polo-box domain (PBD) and prevents PLKs interaction with phosphopeptide substrates

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana J Narvaez et al.
Cell chemical biology, 24(8), 1017-1028 (2017-08-16)
Mutations activating KRAS underlie many forms of cancer, but are refractory to therapeutic targeting. Here, we develop Poloppin, an inhibitor of protein-protein interactions via the Polo-box domain (PBD) of the mitotic Polo-like kinases (PLKs), in monotherapeutic and combination strategies to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service