Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2138

Sigma-Aldrich

IR415

≥98% (HPLC)

Synonym(s):

N-(2,4-Difluorophenyl)-N′-[3-(1H-imidazol-1-yl)propyl]-thiourea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$84.83
25 MG
$360.00

$84.83

List Price$89.30Save 5%
Web-Only Promotion

Available to ship onApril 28, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$84.83
25 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C13H14F2N4S
CAS Number:
Molecular Weight:
296.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$84.83

List Price$89.30Save 5%
Web-Only Promotion

Available to ship onApril 28, 2025Details


Request a Bulk Order

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S=C(NC1=C(F)C=C(F)C=C1)NCCCN2C=NC=C2

InChI

1S/C13H14F2N4S/c14-10-2-3-12(11(15)8-10)18-13(20)17-4-1-6-19-7-5-16-9-19/h2-3,5,7-9H,1,4,6H2,(H2,17,18,20)

InChI key

KRSBPLFMLHNMPU-UHFFFAOYSA-N

Biochem/physiol Actions

HBx is a viral suppressor of host defenses consisting of RNAi-based silencing of viral genes. It is important for HBV replication and also functions as an RNAi suppressor protein. HBx is considered as emerging target for anti-viral therapy.
selective inhibitor of Hepatitis B Virus X Protein (HBx)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhanita Ghosh et al.
The Journal of biological chemistry, 292(30), 12577-12588 (2017-06-07)
Persistent or chronic infection with the hepatitis B virus (HBV) represents one of the most common viral diseases in humans. The hepatitis B virus deploys the hepatitis B virus X protein (HBx) as a suppressor of host defenses consisting of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service