Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2133

Sigma-Aldrich

Ro 65-6570 Hydrochloride

≥98% (HPLC)

Synonym(s):

(RS)-8-(1,2-Dihydro-1-acenaphthylenyl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one Hydrochloride, (RS)-8-Acenaphthen-1-yl-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one hydrochloride, Ro65-6570 Hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$84.83
25 MG
$360.00

$84.83

List Price$89.30Save 5%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$84.83
25 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C25H25N3O · HCl
CAS Number:
Molecular Weight:
419.95
UNSPSC Code:
12352200
NACRES:
NA.77

$84.83

List Price$89.30Save 5%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1NCN(C2=CC=CC=C2)C13CCN(C4CC5=C6C4=CC=CC6=CC=C5)CC3.Cl

InChI key

CTPVGIUCSWWLNA-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

A selective, nonpeptidic, high-affinity nociceptin receptor ORL-1 (ORL1) agonist that elicits anxiolytic-like effects in vivo.
Ro 65-6570 is a nonpeptidic, high-affinity (Ki = 0.52 against 0.1 nM OFQ for binding human ORL1) nociceptin/orphanin FQ (N/OFQ) receptor ORL-1 (KOR-3, NOP) agonist (EC50 = 40 nM by GTPγS binding assay; IC50 = 0.28 nM against 1 μM forskolin-stimulated cellular cAMP accumulation) with good selectivity over opioid receptors μ, k, δ (Ki = 5.9, 26, 250 nM against 1.5 nM naloxone, 3 nM naloxone, 0.3 nM [Ile5,6]-deltorphin II for binding respective receptors), dopamine and serotonin receptors (Ki = 520 nM/D2, 1210 nM/D3,350 nM/D4.4; Ki ≥1 μM for 5HT 1Dα, 2A, 2C, 6, 7). Ro 65-6570 elicits ORL1-dependent anxiolytic-like effects in rats in vivo (0.32-3.2 mg/kg i.p.; elevated plus maze) without affecting spontaneous locomotion. Unlike OFQ, Ro 65-6570 does not affect cocaine-induced conditioned place preference (CPP) in rats.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khaoula Rekik et al.
Neuropharmacology, 125, 39-49 (2017-07-15)
Several neuropeptidergic systems act as modulators of cognitive performances. Among them, nociceptin, an opioid-like peptide also known as orphanin FQ (N/OFQ), has recently gained attention. Stimulation of its receptor, the N/OFQ opioid receptor (NOP), which is expressed in brain regions
A Rizzi et al.
Peptides, 94, 71-77 (2017-07-13)
Nociceptin/orphanin FQ (N/OFQ) modulates several biological functions, including pain transmission via selective activation of a specific receptor named NOP. The aim of this study was the investigation of the antinociceptive properties of NOP agonists and their interaction with opioids in
S Röver et al.
Journal of medicinal chemistry, 43(7), 1329-1338 (2001-02-07)
The discovery of 8-(5,8-dichloro-1,2,3,4-tetrahydro-naphthalen-2-yl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one, 1a, as a high-affinity ligand for the human ORL1 (orphanin FQ/nociceptin) receptor led to the synthesis of a series of optimized ligands. These compounds exhibit high affinity for the human ORL1 receptor, exhibit moderate to good
L Asth et al.
Neuropharmacology, 105, 434-442 (2016-02-13)
Nociceptin/orphanin FQ (N/OFQ) receptor (NOP) agonists produce anxiolytic-like effects in rodents while antagonists promote antidepressant-like effects. The aim of this study was to investigate the effect on anxiety and depression of NOP receptor partial agonists such as the peptides [F/G]N/OFQ(1-13)NH2
Anna Rizzi et al.
Pharmacology research & perspectives, 4(4), e00247-e00247 (2017-01-25)
The aim of the study was to investigate the in vitro and in vivo pharmacological profile of cebranopadol, a novel agonist for opioid and nociceptin/orphanin FQ (N/OFQ) receptors (NOP). In vitro cebranopadol was assayed in calcium mobilization studies in cells coexpressing NOP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service