Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2040

Sigma-Aldrich

Liarozole dihydrochloride

≥98% (HPLC)

Synonym(s):

5-[(3-Chlorophenyl)(1H-imidazol-1-yl)methyl]-1H-benzimidazole, 6-[(3-Chlorophenyl)(1H-imidazol-1-yl)methyl]-1H-benzimidazole dihydrochloride, R 75251 dihydrochloride, R-75251 dihydrochloride, R75251 dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClN4 · 2HCl
CAS Number:
Molecular Weight:
381.69
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=CC(C(C2=CC=C3N=CNC3=C2)N4C=CN=C4)=CC=C1

Biochem/physiol Actions

Liarozole (R 75251, R75,251) is an orally active benzimidazole-based retinoic acid (RA) metabolism blocking agent (RAMBA) that targets multiple P450 enzymes, including aromatase (CYP19), 17-hydroxylase/17,20-lyase (CYP17A1), 11-hydroxylase (CYP11B1), and RA 4-hydroxylase (CYP26). Liarozole is more effective than ketoconazole in vivo (plasma RA enhancement = 2.5 ng/mL vs. 1.3 ng/mL 2 hr after respective oral dosage of 40 mg/kg in rats) and, in contrast to ketoconazole, Liarozole does not significantly affect circulating adrenal androgen levels. Liarozole is shown to effectively suppress androgen-dependent tumor expansion of R3327G Dunning prostate adenocarcinoma xenografts in rats (by 66% and 81% with 80 and 120 mg/kg/day p.o., respectively) independent of its androgen biosynthesis inhibition.
Orally active retinoic acid (RA) metabolism blocking agent (RAMBA) with higher in vivo efficay than ketoconazole.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Van Ginckel et al.
The Prostate, 16(4), 313-323 (1990-01-01)
The antitumoral activity of a novel imidazole derivative, R 75,251, has been studied in the androgen-dependent R3327G Dunning prostate adenocarcinoma grafted subcutaneously in syngeneic rats. Dietary application resulting approximately in dose levels of 80, 120, and 160 mg/kg reduced tumor
J P Van Wauwe et al.
The Journal of pharmacology and experimental therapeutics, 252(1), 365-369 (1990-01-01)
This study examines the effects of ketoconazole, R 75 251 and some other cytochrome P-450 inhibitors on the in vivo metabolism of all-trans-retinoic acid (RA) in normal rats. Oral treatment with ketoconazole or R 75 251 (40 mg/kg, -1 hr)
J Bruynseels et al.
The Prostate, 16(4), 345-357 (1990-01-01)
R 75251, a new imidazole derivative, inhibited the conversion of androgens to estrogens, of progestins to androstenedione and testosterone, and of 11-deoxycorticosterone to corticosterone in human placenta microsomes, subcellular fraction of rat testis, bovine adrenocortical mitochondria, in cultured rat granulosa

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service