Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1934

Sigma-Aldrich

DR4004 hydrochloride

≥98% (HPLC)

Synonym(s):

1-[4-(4-Phenyl-3,6-dihydro-2H-pyridin-1-yl)-butyl]-2a,3,4,5-tetrahydro-1H-benzo[cd]indol-2-one, 2a-[4-(3,6-Dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-2a,3,4,5-tetrahydrobenz[cd]indol-2(1H)-one, DR 4004

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N2O · HCl
CAS Number:
Molecular Weight:
422.99
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

DR4004 is a potent and selective 5-HT7 serotonin receptor antagonist.
Potent and selective 5-HT7 serotonin receptor antagonist

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Takeda et al.
European journal of pharmacology, 518(1), 30-39 (2005-07-09)
The present study examined whether serotonin (5-hydroxytryptamine; 5-HT)7 receptors play a role in the modulation of emotionality in mice using the selective 5-HT7 receptor antagonist 2a-[4-(4-phenyl-1,2,3,6-tetrahydropyridyl)butyl]-2a,3,4,5-tetrahydrobenzo (c,d)indol-2-(1H)-one (DR4004). The emotionality of mice was evaluated in terms of exploratory activity in
Kotaro Takeda et al.
Neuroscience letters, 638, 69-75 (2016-12-13)
Fear conditioning is a valuable behavioral paradigm for studying the neural basis of emotional learning and memory. The present study examined the involvement of extracellular signal-regulated kinase 1/2 (ERK) signaling on the serotonin (5-HT)
Kazumasa Horikawa et al.
Journal of pharmacological sciences, 123(4), 402-406 (2013-12-03)
Several lines of evidence indicate that serotonin type 7 (5-HT7) receptors play a critical role for non-photic resetting of the mammalian circadian clock; however, the contributions of other types of 5-HT receptors to non-photic entrainment are not yet clarified. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service