Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1927

Sigma-Aldrich

LUF7346

≥98% (HPLC)

Synonym(s):

2-[4-(2-Bromobenzoyl)phenoxy]-N-(pyridin-3-yl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$132.05

$132.05

List Price$139.00
Web-Only Promotion

Available to ship onApril 30, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$132.05

About This Item

Empirical Formula (Hill Notation):
C20H15BrN2O3
CAS Number:
Molecular Weight:
411.25
UNSPSC Code:
12352200
NACRES:
NA.77

$132.05

List Price$139.00
Web-Only Promotion

Available to ship onApril 30, 2025Details


Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CN=C1)COC2=CC=C(C(C3=C(Br)C=CC=C3)=O)C=C2

InChI

1S/C20H15BrN2O3/c21-18-6-2-1-5-17(18)20(25)14-7-9-16(10-8-14)26-13-19(24)23-15-4-3-11-22-12-15/h1-12H,13H2,(H,23,24)

InChI key

QNWXOXKHLFTJBA-UHFFFAOYSA-N

Biochem/physiol Actions

LUF7346 has the ability to enhance the potassium current (IKr) in a heterologous system. It is found to be highly effective than other human ether-a-go-go-related gene (hERG) activators, such as Rottlerin and NS1643.[1]
LUF7346 is a Kv11.1 (hERG) allosteric modulator that prevents proarrhythmic effects caused by hERG blockers in rat ventricular myocyte cultures.
LUF7346 is a Kv11.1 (hERG) allosteric modulator that prevents proarrhythmic effects caused by hERG blockers in rat ventricular myocyte cultures. LUF7346 reduces Kv11.1 (ERG) affinity toward known Kv11.1 channel blockers dofetilide (Ki for hERG = 4.8 nM without and 12 nM with 10 μM LUF7346) and astemizole (Ki for for hERG = 1.3 nM without and 5.3 nM with 10 μM LUF7346) and effectively suppresses astemizole-induced arrhythmogenic events in a dose-dependent manner among Long-QT syndrome/LQTS (LQT1, JLNS, LQT2) and control isogenic human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs). LUF7346 is shown to slow IKr deactivation and positively shifts IKr inactivation among LQTS and control isogenic hiPSC-CMs without affecting KCNQ1/KCNE1-dependent IKs or L-type calcium current ICaL.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luca Sala et al.
EMBO molecular medicine, 8(9), 1065-1081 (2016-07-30)
Long-QT syndrome (LQTS) is an arrhythmogenic disorder characterised by prolongation of the QT interval in the electrocardiogram, which can lead to sudden cardiac death. Pharmacological treatments are far from optimal for congenital forms of LQTS, while the acquired form, often
Zhiyi Yu et al.
European journal of medicinal chemistry, 106, 50-59 (2015-11-01)
We synthesized and evaluated a series of compounds for their allosteric modulation at the Kv11.1 (hERG) channel. Most compounds were negative allosteric modulators of [(3)H]dofetilide binding to the channel, in particular 7f, 7h-j and 7p. Compounds 7f and 7p were
A new hERG allosteric modulator rescues genetic and drug-induced long-QT syndrome phenotypes in cardiomyocytes from isogenic pairs of patient induced pluripotent stem cells
Sala L, et al.
EMBO Molecular Medicine, 8(9), 1065-1081 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service