Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1905

Sigma-Aldrich

KR7 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

KPWWPRR-NH2 trifluoroacetate salt, Lys-Pro-Trp-Trp-Pro-Arg-Arg-amide trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H73N17O7 · xC2HF3O2
Molecular Weight:
1024.22 (free base basis)
UNSPSC Code:
12352202

assay

≥98% (HPLC)

form

lyophilized solid

color

white to beige

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

KR7 is a non-toxic and serum stable heptapeptide that inhibits insulin amyloid fibrillation.
KR7 is a non-toxic and serum stable heptapeptide that inhibits insulin amyloid fibrillation. KR7 primarily targets the fiber elongation step.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhisma N Ratha et al.
The Journal of biological chemistry, 291(45), 23545-23556 (2016-09-30)
The aggregation of insulin into amyloid fibers has been a limiting factor in the development of fast acting insulin analogues, creating a demand for excipients that limit aggregation. Despite the potential demand, inhibitors specifically targeting insulin have been few in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service