Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1878

Sigma-Aldrich

GNF-1331

≥98% (HPLC)

Synonym(s):

N-(6-Methoxy-2-benzothiazolyl)-2-[[4-propyl-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N6O2S2
CAS Number:
Molecular Weight:
440.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CCCN1C(SCC(NC2=NC3=C(C=C(OC)C=C3)S2)=O)=NN=C1C4=CC=NC=C4

InChI

1S/C20H20N6O2S2/c1-3-10-26-18(13-6-8-21-9-7-13)24-25-20(26)29-12-17(27)23-19-22-15-5-4-14(28-2)11-16(15)30-19/h4-9,11H,3,10,12H2,1-2H3,(H,22,23,27)

InChI key

JIUFLMSAFMXEEC-UHFFFAOYSA-N

Biochem/physiol Actions

GNF-1331 is an orally available, potent and selective Porcupine (PORCN) inhibitor.
GNF-1331 is an orally available, potent and selective Porcupine inhibitor.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dai Cheng et al.
ACS medicinal chemistry letters, 7(7), 676-680 (2016-07-21)
Blockade of aberrant Wnt signaling is an attractive therapeutic approach in multiple cancers. We developed and performed a cellular high-throughput screen for inhibitors of Wnt secretion and pathway activation. A lead structure (GNF-1331) was identified from the screen. Further studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service