Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1872

Sigma-Aldrich

ORM-3819

≥98% (HPLC)

Synonym(s):

4-Hydroxy-3-methoxy-2-nitro-benzaldehyde 2-[4-[(4R)-1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazone, L-6-{4-[N′-(4-Hydroxi-3-methoxy-2-nitro-benzylidene)-hydrazino]-phenyl}-5-methyl-4,5-dihydro-2 H-pyridazin-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N5O5
CAS Number:
Molecular Weight:
397.38
UNSPSC Code:
12352202

assay

≥98% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: 30 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C[C@H]1C)NN=C1C(C=C2)=CC=C2NN=CC3=C([N+]([O-])=O)C(OC)=C(O)C=C3

Biochem/physiol Actions

ORM-3819 is a potent inotropic agent that binds to cardiac troponin C (cTnC) and sensitizes heart to Ca2+.
ORM-3819 is a potent inotropic agent that binds to cardiac troponin C (cTnC) and sensitizes heart to Ca2+. Also, ORM-3819 is a selective and potent inhibitor of PDE III. ORM-3819 exhibits positive inotropic effects through Ca2+-sensitization and selective inhibition of PDE III. ORM-3819 was effective in vivo dog model of myocardial stunning.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

László Nagy et al.
European journal of pharmacology, 775, 120-129 (2016-02-14)
This study is the first pharmacological characterization of the novel chemical entity, ORM-3819 (L-6-{4-[N'-(4-Hydroxi-3-methoxy-2-nitro-benzylidene)-hydrazino]-phenyl}-5-methyl-4,5-dihydro-2H-pyridazin-3-one), focusing primarily on its cardiotonic effects. ORM-3819 binding to cardiac troponin C (cTnC) was confirmed by nuclear magnetic resonance spectroscopy, and a selective inhibition of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service