Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1850

Sigma-Aldrich

Inz-1

≥98% (HPLC)

Synonym(s):

3-Phenyl-1H-indazole-1-acetic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OC)CN1C2=CC=CC=C2C(C3=CC=CC=C3)=N1

Biochem/physiol Actions

Inz-1 is a potent and selective inhibitor of yeast mitochondrial cytochrome bc1 that reverses resistance to triazole antifungals in the pathogenic fungus Candida albicans.
Potent and selective inhibitor of yeast mitochondrial cytochrome bc1

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willmen Youngsaye et al.
Beilstein journal of organic chemistry, 9, 1501-1507 (2013-08-16)
The National Institutes of Health Molecular Libraries and Probe Production Centers Network (NIH-MLPCN) screened >300,000 compounds to evaluate their ability to restore fluconazole susceptibility in resistant Candida albicans isolates. Additional counter screens were incorporated to remove substances inherently toxic to
Benjamin M Vincent et al.
Cell chemical biology, 23(8), 978-991 (2016-08-16)
To cause disease, a microbial pathogen must adapt to the challenges of its host environment. The leading fungal pathogen Candida albicans colonizes nutrient-poor bodily niches, withstands attack from the immune system, and tolerates treatment with azole antifungals, often evolving resistance.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service