Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1823

Sigma-Aldrich

Vosaroxin hydrochloride

≥98% (HPLC)

Synonym(s):

1,4-Dihydro-7-[(3S,4S)-3-methoxy-4-(methylamino)-1-pyrrolidinyl]-4-oxo-1-(2-thiazolyl)-1,8-naphthyridine-3-carboxylic acid hydrochloride, SNS-595 hydrochloride, Voreloxin hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$612.00

$612.00


Available to ship onApril 29, 2025Details



Select a Size

Change View
25 MG
$612.00

About This Item

Empirical Formula (Hill Notation):
C18H19N5O4S · HCl
CAS Number:
Molecular Weight:
437.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$612.00


Available to ship onApril 29, 2025Details


Quality Level

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +42 to +52°, c = 1 in 0.1 M NaOH

color

white to beige

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O=C(C(C(O)=O)=C1)C2=C(N=C(C=C2)N3C[C@@H]([C@H](C3)NC)OC)N1C4=NC=CS4

InChI

1S/C18H19N5O4S.ClH/c1-19-12-8-22(9-13(12)27-2)14-4-3-10-15(24)11(17(25)26)7-23(16(10)21-14)18-20-5-6-28-18;/h3-7,12-13,19H,8-9H2,1-2H3,(H,25,26);1H/t12-,13-;/m0./s1

InChI key

JJZCCQHWCOXGCL-QNTKWALQSA-N

Biochem/physiol Actions

Vosaroxin is an anti-cancer quinolone derivative that acts as a DNA intercalator and a topoisomerase II inhibitor, causing p53 independent tumor cell apoptosis. Vosaroxin stabilizes cleavage complexes formed by topoisomerase IIα and IIβ, which results in an accumulation of DNA double-strand breaks. Vosaroxin showed antineoplastic activity in multiple solid tumor and hematologic cancer cell lines and has been in clinical trials for AML.
Vosaroxin is an anti-cancer quinolone derivative that acts as a DNA intercalator and a topoisomerase II inhibitor.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas J Short et al.
Expert review of hematology, 9(6), 529-534 (2016-05-10)
The outcomes of patients with relapsed or refractory acute myeloid leukemia (AML) are dismal, and effective treatment options in this patient population are therefore desperately needed. Vosaroxin is a first-in-class anticancer quinolone derivative that has shown promising activity in patients
Gene C Jamieson et al.
Drugs, 76(13), 1245-1255 (2016-08-04)
Vosaroxin is a first-in-class anticancer quinolone derivative that targets topoisomerase II and induces site-selective double-strand breaks in DNA, leading to tumor cell apoptosis. Vosaroxin has chemical and pharmacologic characteristics distinct from other topoisomerase II inhibitors due to its quinolone scaffold.
Christopher B Benton et al.
Future oncology (London, England), 13(2), 125-133 (2016-09-13)
Vosaroxin is a quinolone compound that intercalates DNA and induces TP53-independent apoptosis, demonstrating activity against acute myeloid leukemia (AML) in Phase I-III trials. Here, we examine vosaroxin's mechanism of action and pharmacology, and we review its use in AML to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service