Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1793

Sigma-Aldrich

MI-4F

≥98% (HPLC)

Synonym(s):

2-[2-(4-Fluorophenyl)ethenyl]-3-phenyl-5-(phenylamino)-1,3,4-thiadiazolium chloride, M1 4F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClFN3S
CAS Number:
Molecular Weight:
409.91
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

, light yellow to yellow-orange

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(C=C1)=CC=C1C=CC2=[N+](C3=CC=CC=C3)N=C(NC4=CC=CC=C4)S2.[Cl-]

Biochem/physiol Actions

MI-4F is mesoionic compound that exhibits potent anticancer activity against multiple cancer cell lines and in vivo.
MI-4F is mesoionic compound that exhibits potent anticancer activity against multiple cancer cell lines and in vivo. MI-4F induces apoptosis in HepG2 cells. MI-4F appears to inhibit calcium efflux through inhibition formation of mitochondrial permeability transition pore (MPTP). MI-4F inhibits superoxide dismutase. MI-4F is not susceptible to MDR transporters P-glycoprotein, ABCG2 and MRP1. MI-4F also inhibits ABCG2-mediated efflux of mitoxantrone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda do Rocio Andrade Pires et al.
Chemico-biological interactions, 186(1), 1-8 (2010-04-14)
The main goal of this work was to investigate the relationship between the effects of three new 1,3,4-thiadiazolium mesoionic derivatives on mitochondrial bioenergetics and their previously described chemical structure and antimelanoma activity. The 4-phenyl-5-(2'-Y, 4'-X or 4'-X-cinnamoyl)-1,3,4-thiadiazolium-2-phenylamine chlorides differed from
Gustavo Jabor Gozzi et al.
PloS one, 10(6), e0130046-e0130046 (2015-06-18)
In this work, we evaluated the cytotoxicity of mesoionic 4-phenyl-5-(2-Y, 4-X or 4-X-cinnamoyl)-1,3,4-thiadiazolium-2-phenylamine chloride derivatives (MI-J: X=OH, Y=H; MI-D: X=NO2, Y=H; MI-4F: X=F, Y=H; MI-2,4diF: X=Y=F) on human hepatocellular carcinoma (HepG2), and non-tumor cells (rat hepatocytes) for comparison. MI-J, M-4F
Amanda do Rocio Andrade Pires et al.
European journal of pharmacology, 770, 78-84 (2015-12-17)
Mesoionic compounds have shown antitumor and citotoxic activity against different tumor cells lines, which has been attributed to their physical and chemical characteristics. Among these compounds, the 1,3,4-thiadiazolium-2-phenylamine derivatives have been highlighted due to their important anti-melanoma activity. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service