Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1780

Sigma-Aldrich

BAY-588

≥98% (HPLC)

Synonym(s):

N4-[1-[[4-(1,1-Dimethylethyl)phenyl]methyl]-5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-7-fluoro-2,4-quinolinedicarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25F4N5O2
CAS Number:
Molecular Weight:
527.51
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C2C(N=C(C(N)=O)C=C2C(NC3=C(C)N(CC4=CC=C(C(C)(C)C)C=C4)N=C3C(F)(F)F)=O)=C1

Biochem/physiol Actions

BAY-588 is an inactive control probe for BAY-876. BAY-876 is a potent, highly selective, cell-permeable inhibitor of glucose transporter GLUT1. For characterization details of BAY-876 and BAY-588, please visit the BAY-876 probe summary on the Structural Genomics Consortium (SGC) website.

BAY-876, the active GLUT1 probe, is available from Sigma. To learn more about and purchase BAY-876, click here.

To learn about other SGC chemical probes for protein targets, visit sigma.com/sgc
BAY-588 is an inactive control probe for BAY-876, a potent, highly selective, cell-permeable inhibitor of glucose transporter GLUT1.

Features and Benefits

BAY-588 is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC.
This compound is a featured product for Nitric Oxide & Cell Stress research. Click here to discover more featured Nitric Oxide & Cell Stress products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service