Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1758

Sigma-Aldrich

Tetrazanbigen

≥98% (HPLC)

Synonym(s):

5,6,13,14-Tetrahydro-4bH-6,7,12,12b-tetraaza-benzo[b]chrysene, TNBG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N4
CAS Number:
Molecular Weight:
288.35
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

C12=CC=CC=C1N=C3C(NCC4N3CCC5=C4C=CC=C5)=N2

Biochem/physiol Actions

Tetrazanbigen (TNBG) is a potent and selective antitumor drug that induces lipid accumulation in cancer cells.
Tetrazanbigen (TNBG) is a potent and selective antitumor drug that induces lipid accumulation in cancer cells. Tetrazanbigen induces S phase arrest and apoptosis in hepatocellular carcinoma QGY-7701. Tetrazanbigen potently inhibits growth of QGY-7701 cells xenografts in mice.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service