Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1755

Sigma-Aldrich

ML264

≥98% (HPLC)

Synonym(s):

(2E)-3-(3-Chlorophenyl)-N-[2-[methyl(tetrahydro-1,1-dioxido-2H-thiopyran-4-yl)amino]-2-oxoethyl]-2-propenamide, CID 51003603, SID 117686865, SR03000002171

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21ClN2O4S
CAS Number:
Molecular Weight:
384.88
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mL/mL, clear

Storage temp.

2-8°C

SMILES string

O=C(NCC(N(C)C1CCS(CC1)(=O)=O)=O)/C=C/C2=CC=CC(Cl)=C2

Biochem/physiol Actions

ML264 is a potent and selective inhibitor of Kruppel-like factor 5 (KLF5) expression. ML264 potently inhibits proliferation of colorectal cancer cells in vitro through modifications of the cell-cycle profile.
Potent and selective inhibitor of Kruppel-like factor 5 (KLF5) expression

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ainara Ruiz de Sabando et al.
Molecular cancer therapeutics, 15(1), 72-83 (2015-12-02)
Colorectal cancer is one of the leading causes of cancer mortality in Western civilization. Studies have shown that colorectal cancer arises as a consequence of the modification of genes that regulate important cellular functions. Deregulation of the WNT and RAS/MAPK/PI3K

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service