Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1730

Sigma-Aldrich

LP117

≥98% (HPLC)

Synonym(s):

2-[[4-Chloro-6-(6-quinolinylamino)-2-pyrimidinyl]thio]-hexanoic acid ethyl ester, Ethyl 2-((4-chloro-6-(quinolin-6-ylamino)pyrimidin-2-yl)thio)hexanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClN4O2S
CAS Number:
Molecular Weight:
430.95
UNSPSC Code:
12352202

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=NC(SC(C(OCC)=O)CCCC)=NC(NC2=CC3=C(C=C2)N=CC=C3)=C1

Biochem/physiol Actions

LP117 is a pirinixic acid derivative that inhibits ABCB1-mediated drug transport and resistance in cancer cultures.
LP117 is a pirinixic acid derivative that interferes with ATP-binding cassette (ABC) transporter ABCB1-mediated substrates transport, including vincristine, vinorelbine, paclitaxel, and actinomycin D, but not doxorubicin, rhodamine 123, or JC-1. LP117 effectively reduces drug-resistance in ABCB1-expressing neuroblastoma cultures while exhibiting no efficacy toward non-ABCB1 substrate cisplatin or in non-ABCB1-expressing cancer cultures.

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service