Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1717

Sigma-Aldrich

RA-2

≥98% (HPLC)

Synonym(s):

1,1′-[1,3-Phenylenebis(methylene)]-3-fluoro-4-hydroxy-benzoic acid ester, 1,3-Phenylenebis(methylene)bis(3-fluoro-4-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16F2O6
CAS Number:
Molecular Weight:
414.36
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(O)C(F)=C1)OCC2=CC=CC(COC(C3=CC(F)=C(O)C=C3)=O)=C2

Biochem/physiol Actions

RA-2 is a selective pan-negative-gating modulator of KCa2/3 channels that potently inhibits both ex vivo and in vivo human KCa3.1 and all three human KCa2 channels subtypes. RA-2 right-shifts the KCa3.1 concentration-response curve for Ca2+-activation.
RA-2 is a selective pan-negative-gating modulator of KCa2/3 channels.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service