Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1714

Sigma-Aldrich

DCAI

≥95% (HPLC)

Synonym(s):

2-(4,6-Dichloro-2-methyl-1h-indol-3-yl)ethanamine, 4,6-Dichloro-2-methyl-3-aminoethylindole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$99.50
25 MG
$405.00

$99.50


Check Cart for Availability


Select a Size

Change View
5 MG
$99.50
25 MG
$405.00

About This Item

Empirical Formula (Hill Notation):
C11H12Cl2N2
CAS Number:
Molecular Weight:
243.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$99.50


Check Cart for Availability

Quality Level

assay

≥95% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1c2c([nH]c(c2CCN)C)cc(c1)Cl

InChI

1S/C11H12Cl2N2/c1-6-8(2-3-14)11-9(13)4-7(12)5-10(11)15-6/h4-5,15H,2-3,14H2,1H3

InChI key

JCTJISIFGZHOFY-UHFFFAOYSA-N

Biochem/physiol Actions

DCAI, an Inactive Ras, is bound to a GDP and activated by SOS (son of sevenless, among others), which converts it to the active GTP form. DCAI is a known binder to Ras, which inhibits SOS nucleotide exchange, inhibiting Ras activation.
DCAI, an inactive Ras, is bound to a GDP and activated by SOS (son of sevenless, among others) which converts it to the active GTP form.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jon J G Winter et al.
Journal of medicinal chemistry, 58(5), 2265-2274 (2015-02-20)
Constitutively active mutant KRas displays a reduced rate of GTP hydrolysis via both intrinsic and GTPase-activating protein-catalyzed mechanisms, resulting in the perpetual activation of Ras pathways. We describe a fragment screening campaign using X-ray crystallography that led to the discovery
Thi B Trinh et al.
ACS combinatorial science, 18(1), 75-85 (2015-12-10)
Cyclic peptides have great potential as therapeutic agents and research tools. However, their applications against intracellular targets have been limited, because cyclic peptides are generally impermeable to the cell membrane. It was previously shown that fusion of cyclic peptides with
Till Maurer et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(14), 5299-5304 (2012-03-21)
The Ras gene is frequently mutated in cancer, and mutant Ras drives tumorigenesis. Although Ras is a central oncogene, small molecules that bind to Ras in a well-defined manner and exert inhibitory effects have not been uncovered to date. Through

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service