Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1698

Sigma-Aldrich

TMPPAA

≥98% (HPLC)

Synonym(s):

trans-3-(4-Methoxyphenyl)-N-(pentan-3-yl)acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO2
CAS Number:
Molecular Weight:
247.33
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCC(CC)NC(/C=C/C1=CC=C(OC)C=C1)=O

InChI

1S/C15H21NO2/c1-4-13(5-2)16-15(17)11-8-12-6-9-14(18-3)10-7-12/h6-11,13H,4-5H2,1-3H3,(H,16,17)/b11-8+

InChI key

SPXCFGAFVLQEJK-DHZHZOJOSA-N

Biochem/physiol Actions

TMPPAA is an allosteric agonist and positive allosteric modulator (PAM) of 5-HT3 receptor.
TMPPAA is an allosteric agonist and positive allosteric modulator (PAM) of 5-HT3 receptor. TMPPAA behaves as both an agonist and a PAM at 5-HT3A receptors, not via binding at the orthosteric site.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service