Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1696

Sigma-Aldrich

HPB

≥98% (HPLC)

Synonym(s):

N-Hydroxy-4-((N-(2-hydroxyethyl)-2-phenylacetamido)methyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20N2O4
CAS Number:
Molecular Weight:
328.36
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(N(CCO)CC1=CC=C(C(NO)=O)C=C1)CC2=CC=CC=C2

Biochem/physiol Actions

HPB is a cell-permeable, non-cytotoxic N-hydroxybenzamide derivative that acts as a HDAC6-selective deacetylase inhibitor (IC50 = 31/376/677/842/1133 nM against HDAC6/8/10/1/7, >2 μM against HDAC2/3/4/11/9/5). HPB treatment results in reduced growth, but not viability (up to 64 μM and 72 hrs), of LNCaP prostate cancer cells and non-cancer human foreskin fibroblasts (IC50 = 8 and 16-32 μM, respectively), causing upregulated acetylation levels of alpha-tubulin and peroxiredoxin, but not histones, both in cultures and in mice in vivo. Intraperitoneal injection is efficacious in suppressing the expansion of prostate cancer CWR22 xenograft-derived tumors in mice (by 73% in 4 wks; Five daily 300 mg/kg i.p. dosings a week) without detectable weight loss.
HPB is a cell-permeable, non-cytotoxic N-hydroxybenzamide derivative that acts as a HDAC6-selective deacetylase inhibitor.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service