Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1683

Sigma-Aldrich

MAY0132

≥98% (HPLC)

Synonym(s):

N-(4-Chloro-3-(trifluoromethyl)phenyl)-2,4,6-trimethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15ClF3N
CAS Number:
Molecular Weight:
313.75
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(NC2=CC=C(Cl)C(C(F)(F)F)=C2)C(C)=CC(C)=C1

InChI

1S/C16H15ClF3N/c1-9-6-10(2)15(11(3)7-9)21-12-4-5-14(17)13(8-12)16(18,19)20/h4-8,21H,1-3H3

InChI key

FLFWWGQQNBDVES-UHFFFAOYSA-N

Biochem/physiol Actions

MAY0132 is a selective inhibitor of the cAMP-binding Protein EPAC2 (Exchange Protein directly Activated by Cyclic AMP) subtype 2.
MAY0132 is a selective inhibitor of the cAMP-binding Protein EPAC2 (Exchange Protein directly Activated by Cyclic AMP) subtype 2. MAY0132 has a submicromolar IC50 value at EPAC2, and no activity at EPAC1. MAY0132 is believed to act at an allosteric site, perhaps at the interface of both cAMP binding domains in the EPAC2 protein.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service