Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1676

Sigma-Aldrich

R9-caPep trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

D-Arginyl-D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-arginyl-L-cysteinyl-L-cysteinyl-L-leucylglycyl-L-isoleucyl-L-prolyl-L-α-glutamyl-L-glutaminyl-L-α-glutamyl-L-tyrosine, H-RDRDRDRDRDRDRDRDRD-CC-LGIPEQEY-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C103H183N47O26S2 · xC2HF3O2
Molecular Weight:
2559.98 (free base basis)
UNSPSC Code:
12352202

assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

R9-caPep is an octadecapeptide composed of N-terminal membrane-permeant nona-D-arginine sequence linked via a -Cys-Cys- spacer to a C-terminal proliferating cell nuclear antigen (PCNA) interdomain loop-derived sequence (L126-Y133) that mediates PCNA interaction with its binding partners via their PIP-box domains.
R9-caPep is an octadecapeptide composed of N-terminal membrane-permeant nona-D-arginine sequence linked via a -Cys-Cys- spacer to a C-terminal proliferating cell nuclear antigen (PCNA) interdomain loop-derived sequence (L126-Y133) that mediates PCNA interaction with its binding partners via their PIP-box domains. R9-caPep selectively kills human neuroblastoma cells (IC50 = 10 - 32 μM vs. IC50 = 98 μM/PBMCs and >100 μM/neural crest stem cells), especially those with MYCN gene amplification, by interfering with PCNA-dependent replication fork extension and DSB HR repair, resulting in exacerbated replication stress. R9-caPep intratumoral injection (3 times a week) is reported to suppress SK-N-BE(2)c xenograft tumor expansion in mice in vivo.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service