Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1675

Sigma-Aldrich

SKY Peptide trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

L-Arginyl-L-arginyl-L-tyrosylglycyl-L-threonyl-L-seryl-L-lysyl-L-tyrosyl-L-glutamine TFA salt, RRYGTSKYQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H79N17O15 · xC2HF3O2
CAS Number:
Molecular Weight:
1158.27 (free base basis)
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

SKY Peptide is a non-cytotoxic and stable nonapeptide modeled after human neutrophil peptide 1 (HNP1, a-defensin) β strand 2 CCL5-binding mode to optimally complete against HNP1 for CCL5 interaction (EC50 ~ 0.1 μg/mL against 10 μg/mL HNP1 and 1 μg/mL CCL5 heteromer formation on HUVECs) without affecting CCL5 and cell surface CCR5 interaction. SKY Peptide is shown to inhibit HNP1-CCL5 heteromer-dependent classical monocytes adhesion both in cultures in vitro (EC50 <1 μg/mL; 10 μg/mL HNP1 and 1 μg/mL CCL5 over HUVEC monolayer) and in mice in vivo.
SKY Peptide is a non-cytotoxic and stable nonapeptide.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service