Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1660

Sigma-Aldrich

NA-17

≥98% (HPLC)

Synonym(s):

2-[2-(1,3-Benzodioxol-5-yl)ethyl]-6-[[3-(dimethylamino)propyl]amino]-1H-benz[de]isoquinoline-1,3(2H)-dione, 6-(1,3-Propanediamine-N,N-dimethyl-N’-)-N’’-(3,4-methylenedioxy-phenethylamin)-1,8-naphthalimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27N3O4
CAS Number:
Molecular Weight:
445.51
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

light yellow to orange

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN(C)CCCNC1=CC=C(C(N(CCC2=CC=C(OCO3)C3=C2)C4=O)=O)C5=C4C=CC=C51

InChI

1S/C26H27N3O4/c1-28(2)13-4-12-27-21-9-8-20-24-18(21)5-3-6-19(24)25(30)29(26(20)31)14-11-17-7-10-22-23(15-17)33-16-32-22/h3,5-10,15,27H,4,11-14,16H2,1-2H3

InChI key

ACOYDEZACFFCPN-UHFFFAOYSA-N

Biochem/physiol Actions

NA-17 is a fluorescent naphthalimide that induces accumulation of activated p53 in mitochondria and nuclei of NSCLC cells.
NA-17 is a fluorescent naphthalimide that induces accumulation of activated p53 in mitochondria and nuclei of NSCLC cells. NA-17 reorganizes the Bak–Bcl-xl complex, promotes p53 phosphorylation without altering the expression of p53, induces cell cycle arrest at the G1 phase and apoptosis. NA-17 exhibits potent anticancer activities in multiple cancer cell lines showing low toxicity to normal cell lines.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service